missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Butoxyethoxy)ethanol, TRC

Product Code. 30332739
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332739 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332739 Supplier Toronto Research Chemicals Supplier No. TRCB28171510MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylene glycol-monobutyl ether
CAS 112-34-5
Molecular Formula C8 H18 O3
Quantity 10 mg
Synonym 2-(2-Butoxyethoxy)ethanol, 2-(2-n-Butoxyethoxy)ethanol, 3,6-Dioxa-1-decanol, BDG, BDG-NS, BDGE 20, Bikanol B 2, Butadigol, Butoxyethoxyethanol, Butycenol 20P, Butyl Carbitol, Butyl Diglysolv, Butyl Oxitol glycol ether, Butyl diglycol, Butyl digol, Butyl dioxitol, Butysenol 20, Butysenol 20P, C 4E2, Circuposit 3304, Circuposit 4125, Circuposit Hole Prep 3304, Circuposit Hole Prep 4125, DB, Diethylene glycol butyl ether, Diethylene glycol mono-n-butyl ether, Diethylene glycol monobutyl ether, Diethylene glycol n-butyl ether, Diglycol monobutyl ether, Dowanol DB, Ektasolve DB, Ethanol, 2,2'-oxybis-, monobutyl ether, Glycol Ether DB, Hisolve DB, K 50181, NBC, NBC (solvent), NSC 407762, O-Butyl diethylene glycol, Poly-Solv DB, Sinopol BDG-LS, n-Butyl carbitol
InChI Formula InChI=1S/C8H18O3/c1-2-3-5-10-7-8-11-6-4-9/h9H,2-8H2,1H3
SMILES CCCCOCCOCCO
IUPAC Name 2-(2-butoxyethoxy)ethanol
Molecular Weight (g/mol) 162.23
Formula Weight 162.1256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.