Learn More
2-(2-Bromoethyl)-1,3-dioxolane, 95%, stab. with silver
Description
2-(2-Bromoethyl)-1,3-dioxolane is used as a pharmaceutical intermediate, dioxolanes, acetals, ketals, building blocks, chemical synthesis, organic building blocks and oxygen compounds. 1,1-(Ethylenedioxy)-3-bromopropane is used in the synthesis of EGFR inhibitors. Also used in the synthesis of orally active agents of cancer and cell proliferation with quinoline substructures. It is also used as flavouring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-(2-Bromoethyl)-1,3-dioxolane is used as a pharmaceutical intermediate, dioxolanes, acetals, ketals, building blocks, chemical synthesis, organic building blocks and oxygen compounds. 1,1-(Ethylenedioxy)-3-bromopropane is used in the synthesis of EGFR inhibitors. Also used in the synthesis of orally active agents of cancer and cell proliferation with quinoline substructures. It is also used as flavouring agent.
Solubility
It is immiscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive. Store under inert gas.
Specifications
Specifications
| Density | 1.512 |
| Boiling Point | 68°C to 70°C (8 mmHg) |
| Flash Point | 65°C (149°F) |
| Refractive Index | 1.479 |
| Quantity | 10 g |
| UN Number | UN2810 |
| Beilstein | 103516 |
| Sensitivity | Light sensitive |
| Solubility Information | It is immiscible with water. |
| Formula Weight | 181.04 |
| Show More |
Safety and Handling
- 2-(2-Bromoethyl)-1,3-dioxolane
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: 2-(2-Bromoethyl)-1,3-dioxolane
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.