missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoethyl)-1,3-dioxane, 98%

Product Code. 11394384
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394384 10 g 10g
11304394 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394384 Supplier Thermo Scientific Alfa Aesar Supplier No. L00958.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 33884-43-4 | C6H11BrO2 | 195.06 g/mol

2-(2-Bromoethyl)-1,3-dioxane, Provides a useful means of introducing a masked β-formylethyl group. In comparison with the corresponding dioxolane (following entry), the dioxane is more stable to acid, and the Grignard is thermally stable. Reaction of the Grignard with an acid chloride gives the ketone in high yield without further reaction to the tertiary alcohol. Subsequent hydrolysis of the 1,3-dioxane gives the -ketoaldehyde. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Bromoethyl)-1,3-dioxane, Provides a useful means of introducing a masked ß-formylethyl group. In comparison with the corresponding dioxolane (following entry), the dioxane is more stable to acid, and the Grignard is thermally stable. Reaction of the Grignard with an acid chloride gives the ketone in high yield without further reaction to the tertiary alcohol. Subsequent hydrolysis of the 1,3-dioxane gives the -ketoaldehyde. It is also used as a pharmaceutical intermediate.

Solubility
It is not soluble in water but soluble in organic solvent such as ethanol.

Notes
Air, Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33884-43-4
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00006567
InChI Key KXMZOKKPQZRPRN-UHFFFAOYNA-N
Synonym 2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane
PubChem CID 520656
SMILES CC(Br)C1OCCCO1

Specifications

Density 1.434
Boiling Point 67°C to 70°C (3 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.482
Quantity 10 g
Beilstein 1421628
Solubility Information It is not soluble in water but soluble in organic solvent such as ethanol.
IUPAC Name 2-(2-bromoethyl)-1,3-dioxane
Formula Weight 195.06
Percent Purity 98%
Chemical Name or Material 2-(2-Bromoethyl)-1,3-dioxane
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.