missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoethoxy)tetrahydro-2H-pyran, 96%, stabilized

Product Code. 10245024 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245024 25 mL Glass bottle 25mL
10021234 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245024 Supplier Thermo Scientific Acros Supplier No. 394930250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17739-45-6 | C7H13BrO2 | 209.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17739-45-6
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD01321310
InChI Key GCUOLJOTJRUDIZ-UHFFFAOYSA-N
Synonym 2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran
PubChem CID 86621
IUPAC Name 2-(2-bromoethoxy)oxane
SMILES C1CCOC(C1)OCCBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 1.3840g/mL
Boiling Point 62°C to 64°C (0.4 mmHg)
Flash Point 87°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 25 mL
Specific Gravity 1.384
Solubility Information Solubility in water: insoluble
Formula Weight 209.08
Percent Purity 96%
Chemical Name or Material 2-(2-Bromoethoxy)tetrahydro-2H-pyran
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Bromoethoxy)tetrahydro-2H-pyran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.