missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoacetyl)thiazole, 97%, Thermo Scientific™

Product Code. 15209736 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15209736 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15209736 Supplier Thermo Scientific Acros Supplier No. 451470010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3292-77-1
Molecular Formula C5H4BrNOS
Molecular Weight (g/mol) 206.06
MDL Number MFCD06411540
InChI Key AQRFTRDAOYSMEA-UHFFFAOYSA-N
Synonym 2-bromo-1-thiazol-2-yl ethanone, 2-bromo-1-1,3-thiazol-2-yl ethanone, 2-bromoacetyl-1,3-thiazole, 2-bromo-1-thiazol-2-yl-ethanone, 2-bromo-1-1,3-thiazol-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thiazolyl, 2-bromo-1-1,3-thiazol-2-yl-1-ethanone, 1-bromo-1,3-thiazole-2-yl ethane-2-one, 2-bromoacetylthiazole, 2-bromoacetyl thiazole
PubChem CID 2795212
IUPAC Name 2-bromo-1-(1,3-thiazol-2-yl)ethanone
SMILES BrCC(=O)C1=NC=CS1

Specifications

Melting Point 52.0°C to 55.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 206.06
Percent Purity 97%
Chemical Name or Material 2-(2-Bromoacetyl)thiazole
compliance-icons compliance-icons
Product Identifier
  • 2-(2-Bromoacetyl)thiazole, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.