missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromo-6-chlorophenyl)acetic Acid, TRC

Product Code. 30330924
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 30330924

Brand: Toronto Research Chemicals TRCB59429050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-Bromo-6-chlorophenyl)acetic Acid
CAS 37777-77-8
Molecular Formula C8H6O2ClBr
Quantity 50 mg
Synonym 2-Chloro-6-bromophenylacetic Acid, 2-Bromo-6-chlorobenzeneacetic Acid
InChI Formula InChI=1S/C8H6BrClO2/c9-6-2-1-3-7(10)5(6)4-8(11)12/h1-3H,4H2,(H,11,12)
SMILES OC(=O)Cc1c(Cl)cccc1Br
IUPAC Name 2-(2-bromo-6-chlorophenyl)acetic acid
Molecular Weight (g/mol) 249.49
Formula Weight 247.924
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.