missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(hydroxymethyl)propionic Acid, TRC
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
Specifications
Specifications
| Chemical Name or Material | 2,2-Dimethylolpropionic acid |
| CAS | 4767-03-7 |
| Molecular Formula | C5 H10 O4 |
| Quantity | 50 g |
| Synonym | Propionic acid, 2,2-bis(hydroxymethyl)- (6CI,7CI,8CI), 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid, 2,2-Bis(hydroxymethyl)propanoic acid, 2,2-Bis(hydroxymethyl)propionic acid, 2,2-Bis(methylol)propionic acid, 2,2-Dihydroxymethylpropanoic acid, 2,2-Dimethylolpropionic acid, 3-Hydroxy-2-hydroxymethyl-2-methylpropionic acid, Bis(hydroxymethyl)propionic acid, Bis-MPA, DMPA, Dimethylolpropionic acid, NSC 96616, Nikkamer PA, Propanoic acid, 2,2-bis(hydroxymethyl)-, α,α-Bis(hydroxymethyl)propionic acid, α,α-Dimethylolpropionic acid, 2,2-Dimethylolpropionic Acid |
| InChI Formula | InChI=1S/C5H10O4/c1-5(2-6,3-7)4(8)9/h6-7H,2-3H2,1H3,(H,8,9) |
| SMILES | CC(CO)(CO)C(=O)O |
| IUPAC Name | 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid |
| Molecular Weight (g/mol) | 134.1305 |
| Formula Weight | 134.0579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction