missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(hydroxymethyl)propionic Acid, TRC

Product Code. 30332924
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332924 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332924 Supplier Toronto Research Chemicals Supplier No. TRCB44744825G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Dimethylolpropionic acid
CAS 4767-03-7
Molecular Formula C5 H10 O4
Quantity 25 g
Synonym Propionic acid, 2,2-bis(hydroxymethyl)- (6CI,7CI,8CI), 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid, 2,2-Bis(hydroxymethyl)propanoic acid, 2,2-Bis(hydroxymethyl)propionic acid, 2,2-Bis(methylol)propionic acid, 2,2-Dihydroxymethylpropanoic acid, 2,2-Dimethylolpropionic acid, 3-Hydroxy-2-hydroxymethyl-2-methylpropionic acid, Bis(hydroxymethyl)propionic acid, Bis-MPA, DMPA, Dimethylolpropionic acid, NSC 96616, Nikkamer PA, Propanoic acid, 2,2-bis(hydroxymethyl)-, α,α-Bis(hydroxymethyl)propionic acid, α,α-Dimethylolpropionic acid, 2,2-Dimethylolpropionic Acid
InChI Formula InChI=1S/C5H10O4/c1-5(2-6,3-7)4(8)9/h6-7H,2-3H2,1H3,(H,8,9)
SMILES CC(CO)(CO)C(=O)O
IUPAC Name 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
Molecular Weight (g/mol) 134.1305
Formula Weight 134.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.