missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene]chloro(p-cymene)ruthenium Chloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | [2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene]chloro(p-cymene)ruthenium Chloride |
| CAS | 130004-33-0 |
| Molecular Formula | C45 H35 Cl P2 Ru . C10 H20 . Cl |
| Quantity | 500 mg |
| Synonym | [1,1'-(1S)-[1,1'-Binaphthalene]-2,2'-diylbis[1,1-diphenylphosphine-κP]]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium(1+) Chloride, (S)-[[1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine]-P,P']chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium(1+) Chloride, (S)-[1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine Ruthenium Complex, ((S)-(2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl))(chloro)(η6-p-isopropyltoluene)ruthenium(1+) Chloride, ((S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl)(chloro)(η6-4-isopropyltoluene)ruthenium(1+) Chloride, [(1S)-[1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine-κP]]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium(1+) Chloride |
| InChI Formula | InChI=1S/C44H32P2.C10H20.CH3.2ClH.Ru/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;1-8(2)10-6-4-9(3)5-7-10;;;;/h1-32H;8-10H,4-7H2,1-3H3;1H3;2*1H;/q;;;;;+2/p-2/t;9-,10+;;;; |
| SMILES | [Cl-].[Cl-][Ru+2]1(C)[P](c2ccccc2)(c3ccccc3)c4ccc5ccccc5c4c6c(ccc7ccccc67)[P]1(c8ccccc8)c9ccccc9.CC(C)[C@@H]%10CC[C@H](C)CC%10 |
| IUPAC Name | dichlororuthenium;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene |
| Molecular Weight (g/mol) | 949.949 |
| Formula Weight | 949.22 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction