missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis-(4-hydroxyphenyl)hexafluoropropane, TRC

Product Code. 18092800
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18092800 5 g 5g
17949819 1 g 1g
18000510 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18092800 Supplier Toronto Research Chemicals Supplier No. TRCB4472905G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1478-61-1
Molecular Formula C15 H10 F6 O2
Molecular Weight (g/mol) 336.23
Synonym Phenol, 4,4'-[2,2,2-trifluoro-1-(trifluoromethyl)ethylidene]di- (6CI,7CI), Phenol, 4,4'-[trifluoro-1-(trifluoromethyl)ethylidene]di- (8CI), 4,4'-[2,2,2-Trifluoro-1-(trifluoromethyl)ethylidene]bis[phenol], 1,1,1,3,3,3-Hexafluoro-2,2-bis(4-hydroxyphenyl)propane, 2,2-(4-Hydroxyphenyl)hexafluoropropane, 2,2-Bis(4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropane, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, 2,2-Bis(4-hydroxyphenyl)perfluoropropane, 2,2-Bis(4'-hydroxyphenyl)hexafluoropropane, 2,2-Bis(p-hydroxyphenyl)hexafluoropropane, 4,4'-(Hexafluoroisopropylidene)bisphenol, 4,4'-(Hexafluoroisopropylidene)diphenol, AF 50, BIS-AF, BIS-AF(H), Biphenol AF, Bisphenol AE, Bisphenol AF, Cheminox BAF, Curative 30, GP 21, Hexafluorobisphenol A, Hexafluorodiphenylolpropane, Hexafluoroisopropylidenebis(4-hydroxybenzene), NSC 152522
IUPAC Name 4-[1,1,1,3,3,3-hexafluoro-2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES Oc1ccc(cc1)C(c2ccc(O)cc2)(C(F)(F)F)C(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H10F6O2/c16-14(17,18)13(15(19,20)21,9-1-5-11(22)6-2-9)10-3-7-12(23)8-4-10/h1-8,22-23H
Formula Weight 336.0585
Chemical Name or Material Bisphenol AF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.