missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%

Product Code. 15471178
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471178 25 g 25g
15481178 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471178 Supplier Thermo Scientific Alfa Aesar Supplier No. H55057.14
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 13080-86-9 | C27H26N2O2 | 410.517 g/mol

It is applied in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in pharmaceutical industry.

Solubility
Very slightly soluble in water. Soluble in acetone, Dimethyl sulfoxide(DMSO).

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13080-86-9
Molecular Formula C27H26N2O2
Molecular Weight (g/mol) 410.517
MDL Number MFCD00039152
InChI Key KMKWGXGSGPYISJ-UHFFFAOYSA-N
Synonym 2,2-bis 4-4-aminophenoxy phenyl propane, 4,4'-propane-2,2-diylbis 4,1-phenylene bis oxy dianiline, 2,2'-bis 4-aminophenoxyphenyl propane, benzenamine, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bis, 4,4'-isopropylidenebis 4,1-phenyleneoxy dianiline, 4,4'-4,4'-isopropylidenediphenyl-1,1'-diyldioxy dianiline, bis 4-4-aminophenoxy phenyl dimethyl methane, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bisbenzenamine
PubChem CID 83119
IUPAC Name 4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline
SMILES CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N)C3=CC=C(C=C3)OC4=CC=C(C=C4)N

Specifications

Melting Point 127°C to 130°C
Quantity 25 g
Solubility Information Very slightly soluble in water. Soluble in acetone,Dimethyl sulfoxide(DMSO).
Formula Weight 410.52
Percent Purity 98%
Chemical Name or Material 2,2-Bis[4-(4-aminophenoxy)phenyl]propane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.