missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(3-Amino-4-hydroxyphenyl(hexafluoropropane)), TRC
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Specifications
Specifications
| Chemical Name or Material | 2,2-Bis(3-Amino-4-hydroxyphenyl(hexafluoropropane)) |
| CAS | 83558-87-6 |
| Molecular Formula | C15H12F6N2O2 |
| Quantity | 250 mg |
| Synonym | 1,1-Bis(3-amino-4-hydroxyphenyl)-2,2,2-trifluoro-1-(trifluoromethyl)ethane, 2,2-Bis(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropane, 2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, 2,2'-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, 2-Amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol, 4,4'-(Hexafluoroisopropylidene)bis[2-aminophenol], 4,4'-Dihydroxy-3,3'-diaminophenylhexafluoropropane, BAFA, BAHF, BIS-AP-AF, Bis(aminophenol) AF, Di(aminobisphenol) AF, Hexafluoro-2,2-bis(3-amino-4-hydroxyphenyl)propane |
| InChI Formula | InChI=1S/C15H12F6N2O2/c16-14(17,18)13(15(19,20)21,7-1-3-11(24)9(22)5-7)8-2-4-12(25)10(23)6-8/h1-6,24-25H,22-23H2 |
| SMILES | Nc1cc(ccc1O)C(c2ccc(O)c(N)c2)(C(F)(F)F)C(F)(F)F |
| IUPAC Name | 2-amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol |
| Molecular Weight (g/mol) | 366.26 |
| Formula Weight | 366.0803 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction