missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Biquinoline, 98%

Product Code. 11434283
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434283 1 g 1g
11444283 5 g 5g
11454283 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434283 Supplier Thermo Scientific Alfa Aesar Supplier No. A10907.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-91-5 | C18H12N2 | 256.308 g/mol

2,2'-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.

Solubility
Sparingly soluble in water (0.001g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-91-5
Molecular Formula C18H12N2
Molecular Weight (g/mol) 256.308
MDL Number MFCD00006740
InChI Key WPTCSQBWLUUYDV-UHFFFAOYSA-N
Synonym 2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline
PubChem CID 8412
IUPAC Name 2-quinolin-2-ylquinoline
SMILES C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3

Specifications

Melting Point 193°C to 196°C
Quantity 1 g
Beilstein 187259
Solubility Information Sparingly soluble in water (0.001g/L).
Formula Weight 256.31
Percent Purity ≥98%
Chemical Name or Material 2,2'-Biquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.