missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyrimidine, 96%

Product Code. 11314504
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11314504

Brand: Thermo Scientific Alfa Aesar L01190.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34671-83-5 | C8H6N4 | 158.16 g/mol

2,2'-Bipyrimidine is used in the preparation of 5-bromo-[2,2']bipyrimidinyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Bipyrimidine is used in the preparation of 5-bromo-[2,2′]bipyrimidinyl.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34671-83-5
Molecular Formula C8H6N4
Molecular Weight (g/mol) 158.16
MDL Number MFCD00014600
InChI Key HKOAFLAGUQUJQG-UHFFFAOYSA-N
Synonym 2,2'-bipyrimidine, 2,2'-bipyrimidyl, 2,2-bipyrimidine, bpym, 2-pyrimidin-2-yl pyrimidine, bipyrimidine, 2,2'-bipyrimidinyl, 2,2'-dipyrimidyl, 2,2;-bipyrimidine
PubChem CID 123444
SMILES C1=CN=C(N=C1)C1=NC=CC=N1

Specifications

Melting Point 110°C to 116°C
Quantity 250 mg
Beilstein 607396
Solubility Information Soluble in methanol.
IUPAC Name 2-pyrimidin-2-ylpyrimidine
Formula Weight 158.16
Percent Purity 96%
Chemical Name or Material 2,2'-Bipyrimidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.