missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azobis[2-methyl-propanimidamide] Dihydrochloride, TRC

Product Code. 30342434
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342434 25 g 25g
30341701 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30342434 Supplier Toronto Research Chemicals Supplier No. TRCA93265025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2'-Azobis[2-methyl-propanimidamide] Dihydrochloride
CAS 2997-92-4
Molecular Formula C8H20Cl2N6
Quantity 25 g
Synonym 2,2'-(1,2-Diazenediyl)bis[2-methylpropanimidamide Dihydrochloride, 2,2'-Azobis[2-methyl-propionamidine Dihydrochloride, 2,2-Azobis(2-amidinopropane) Dihydrochloride, 2,2'-Azobis(2-amidinopropane) Dihydrochloride, 2,2'-Azobis(2-methylpropionamidine) Dihydrochloride, 2,2'-Azobis(isobutyramidine hydrochloride), 2,2'-Azobis(isobutyramidine) Dihydrochloride, 2,2'-Azobis(propane-2-carboxamidine) Dihydrochloride, 2,2'-Azobisamidinopropane dihydrochloride, 2,2'-Azobisisobutyramidinium Chloride, AAPH, AIBA, Azobis(isobutyramidine) Dihydrochloride, Azobisisobutyramidinium Dichloride, Azostarter V 50, MS 1, MS 1 (catalyst), NC 32P, V 50, VA 50, VA 50 (catalyst), Vazo 56, Vazo 56WSP, Vazo 56WSW, Wako V 50
InChI Formula InChI=1S/C8H18N6.2ClH/c1-7(2,5(9)10)13-14-8(3,4)6(11)12;;/h1-4H3,(H3,9,10)(H3,11,12);2*1H
SMILES Cl.Cl.CC(C)(N=NC(C)(C)C(=N)N)C(=N)N
IUPAC Name 2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride
Molecular Weight (g/mol) 271.19
Formula Weight 270.1127
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.