missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic Acid Solution, 1 liter, MP Biomedicals

Product Code. 11425812 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425812 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11425812 Supplier MP Biomedicals Supplier No. 04821810
  • ONLINE EXCLUSIVE PRICE
    £621.00
    £581.70 / 1 L
    Save £39.30
    6% Off
 Restricted Item
Estimated Shipment: 04-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stabilized Liquid Substrate A safe, convenient and good overall performing substrate for ELISA. It may be used for both kinetic and endpoint reactions; Supplied ready-to-use at 1.46 mmol/liter

Specifications

CAS 30931-67-0
Molecular Formula C18H24N6O6S4
MDL Number MFCD00010404,MFCD00010404
Quantity 1 L
Synonym ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
InChI Key OHDRQQURAXLVGJ-AXMZSLBLSA-N
SMILES [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O
IUPAC Name diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
Molecular Weight (g/mol) 548.67
PubChem CID 91884754
Chemical Name or Material 2, 2'-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic Acid Solution, 1 liter
Show More Show Less
compliance-icons
Product Identifier
  • 2,2'-Azino-bis-(3-ethylbenzthiazoline-6-sulfonic Acid) Solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2,2'-Azino-bis-(3-ethylbenzthiazoline-6-sulfonic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.