missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Anhydrouridine, MedChemExpress

Product Code. 16469356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469356 Supplier Medchem Express Supplier No. HYW012313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2,2'-Anhydrouridine is used for anticancer and antiviral research.

2,2'-Anhydrouridine is used for anticancer and antiviral research.

Specifications

Chemical Name or Material 2,2'-Anhydrouridine
Physical Form Powder
CAS 3736-77-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H10N2O5
For Use With (Application) Cancer-programmed cell death
Synonym 2,2'-Cyclouridine O2,2'-Cyclouridine
Solubility Information DMSO : 25 mg/mL (110.53 mM; Need ultrasonic)
SMILES OC[C@@H](O1)[C@@H](O)[C@@](O2)([H])[C@]1([H])N(C2=N3)C=CC3=O
Molecular Weight (g/mol) 226.19
Formula Weight 226.19
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.