missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminothiazol-4-yl)acetic acid hydrochloride, TRC

Product Code. 30329782
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30329782

Brand: Toronto Research Chemicals TRCA63333350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-Aminothiazol-4-yl)acetic acid hydrochloride
CAS 66659-20-9
Molecular Formula C5H6N2O2S • HCl
Quantity 50 mg
Synonym 4-Thiazoleacetic acid, 2-amino-, hydrochloride (1:1), 4-Thiazoleacetic acid, 2-amino-, monohydrochloride (9CI), 2-(2-Amino-1,3-thiazol-4-yl)acetic acid hydrochloride, 2-Aminothiazole-4-acetic acid hydrochloride
InChI Formula InChI=1S/C5H6N2O2S.ClH/c6-5-7-3(2-10-5)1-4(8)9;/h2H,1H2,(H2,6,7)(H,8,9);1H
SMILES C(C1=CSC(=N)N1)C(=O)O.Cl
IUPAC Name 2-(2-amino-1,3-thiazol-4-yl)acetic acid;hydrochloride
Molecular Weight (g/mol) 194.63
Formula Weight 193.9917
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.