missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Aminothiazol-4-yl)acetic acid hydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2-(2-Aminothiazol-4-yl)acetic acid hydrochloride |
| CAS | 66659-20-9 |
| Molecular Formula | C5H6N2O2S • HCl |
| Quantity | 10 mg |
| Synonym | 4-Thiazoleacetic acid, 2-amino-, hydrochloride (1:1), 4-Thiazoleacetic acid, 2-amino-, monohydrochloride (9CI), 2-(2-Amino-1,3-thiazol-4-yl)acetic acid hydrochloride, 2-Aminothiazole-4-acetic acid hydrochloride |
| InChI Formula | InChI=1S/C5H6N2O2S.ClH/c6-5-7-3(2-10-5)1-4(8)9;/h2H,1H2,(H2,6,7)(H,8,9);1H |
| SMILES | C(C1=CSC(=N)N1)C(=O)O.Cl |
| IUPAC Name | 2-(2-amino-1,3-thiazol-4-yl)acetic acid;hydrochloride |
| Molecular Weight (g/mol) | 194.63 |
| Formula Weight | 193.9917 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction