missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminophenyl)benzimidazole, 98%

Product Code. 11426740
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426740 5 g 5g
11436740 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426740 Supplier Thermo Scientific Alfa Aesar Supplier No. L07034.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5805-39-0 | C13H11N3 | 209.25 g/mol

2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5805-39-0
Molecular Formula C13H11N3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00051496
InChI Key YWNXHTNWOQHFRL-UHFFFAOYSA-N
Synonym 2-2-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl aniline, 2-1h-benzimidazol-2-yl aniline, 2-1h-benzoimidazol-2-yl-phenylamine, 2-1h-1,3-benzodiazol-2-yl aniline, benzenamine, 2-1h-benzimidazol-2-yl, 2-o-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl benzenamine, 2-benzimidazol-2-ylphenylamine
PubChem CID 79869
SMILES NC1=CC=CC=C1C1=NC2=CC=CC=C2N1

Specifications

Melting Point 212°C to 216°C
Quantity 5 g
Beilstein 173018
Solubility Information Insoluble in water.
IUPAC Name 2-(1H-1,3-benzodiazol-2-yl)aniline
Formula Weight 209.24
Percent Purity 98%
Chemical Name or Material 2-(2-Aminophenyl)benzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.