missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethylamino)ethanol, TRC

Product Code. 30133658
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133658 100 g 100g
30012487 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133658 Supplier Toronto Research Chemicals Supplier No. TRCA608990100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-41-1
Molecular Formula C4 H12 N2 O
Molecular Weight (g/mol) 104.1509
Synonym 2-[(2-Aminoethyl)amino]ethanol, (2-Hydroxyethyl)ethylenediamine, (β-Hydroxyethyl)ethylenediamine, 1-(2-Hydroxyethylamino)-2-aminoethane, 1-Aminooxyethylamine, 2-(2-Aminoethylamino)-1-ethanol, 2-(2-Hydroxyethylamino)ethylamine, 2-(2'-Aminoethylamino)ethanol, 2-[(Aminoethyl)amino]ethanol, A-EA, Amino Alcohol EA, Aminoethylethanolamine, N-(2-Aminoethyl)ethanolamine, N-(2-Hydroxyethyl)-1,2-diaminoethane, N-(2-Hydroxyethyl)-1,2-ethanediamine, N-(2-Hydroxyethyl)-1,2-ethylenediamine, N-(2-Hydroxyethyl)ethylenediamine, N-(2'-Hydroxyethyl)ethylenediamine, N-(Hydroxyethyl)ethylenediamine, N-(β-Aminoethyl)ethanolamine, N-(β-Hydroxyethyl)-1,2-ethanediamine, N-(β-Hydroxyethyl)ethylenediamine, N-Hydroxyethyl-1,2-ethanediamine, NSC 461
IUPAC Name 2-(2-aminoethylamino)ethanol
SMILES NCCNCCO

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H12N2O/c5-1-2-6-3-4-7/h6-7H,1-5H2
Formula Weight 104.095
Chemical Name or Material (2-Hydroxyethyl)ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.