missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(2-Aminoethyl)thio]ethan-1-ol, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
CAS Min % | 90.0 |
Density | 1.1200g/mL |
Assay Percent Range | 90% |
Linear Formula | NH2CH2CH2SCH2CH2OH |
Quantity | 5 g |
Formula Weight | 121.2 |
Percent Purity | 90% |
Chemical Name or Material | 2-[(2-Aminoethyl)thio]ethan-1-ol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'