Learn More
2-(2-Aminoethyl)-1-methylpyrrolidine, 97%, Thermo Scientific Chemicals
£35.42 - £122.00
Chemical Identifiers
CAS | 51387-90-7 |
---|---|
Molecular Formula | C7H16N2 |
Molecular Weight (g/mol) | 128.22 |
MDL Number | MFCD00003175 |
InChI Key | PNHGJPJOMCXSKN-UHFFFAOYNA-N |
Synonym | 2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine |
PubChem CID | 98388 |
SMILES | CN1CCCC1CCN |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51387-90-7 | |
128.22 | |
PNHGJPJOMCXSKN-UHFFFAOYNA-N | |
98388 |
C7H16N2 | |
MFCD00003175 | |
2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine | |
CN1CCCC1CCN |
Specifications
51387-90-7 | |
100.0 | |
0.8800g/mL | |
Authentic | |
C7H16N2 | |
MFCD00003175 | |
2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine | |
CN1CCCC1CCN | |
128.22 | |
128.22 | |
Powder |
96.0 | |
Beige to Brown | |
65°C | |
96% min. (GC) | |
1.4684 | |
0.88 | |
PNHGJPJOMCXSKN-UHFFFAOYNA-N | |
2-(1-methylpyrrolidin-2-yl)ethan-1-amine | |
98388 | |
97% | |
2-(2-Aminoethyl)-1-methylpyrrolidine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 257-169-
- 2-(2-Aminoethyl)-1-methylpyrrolidine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only