missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethyl)-1-methylpyrrolidine, 97%, Thermo Scientific Chemicals

Product Code. 10306834 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306834 5 g Glass bottle 5g
10061694 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10306834 Supplier Thermo Scientific Acros Supplier No. 160860050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51387-90-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00003175
InChI Key PNHGJPJOMCXSKN-UHFFFAOYNA-N
Synonym 2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine
PubChem CID 98388
SMILES CN1CCCC1CCN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Beige to Brown
Density 0.8800g/mL
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4684
Quantity 5 g
Specific Gravity 0.88
IUPAC Name 2-(1-methylpyrrolidin-2-yl)ethanamine
Formula Weight 128.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-(2-Aminoethyl)-1-methylpyrrolidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(2-Aminoethyl)-1-methylpyrrolidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.