missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-[(Benzyloxy)carbonyl]-L-valinate, Mikromol™
Description
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-[(Benzyloxy)carbonyl]-L-valinate
Specifications
Specifications
| Chemical Name or Material | 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-[(benzyloxy)carbonyl]-L-valinate |
| CAS | 124832-31-1 |
| Analyte or Component Names | 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-[(benzyloxy)carbonyl]-L-valinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 N6 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Valaciclovir Hydrochloride Imp. E (EP), Valaciclovir Imp. E (EP), Valacyclovir USP Related Compound E, Valacyclovir USP RC E, L-Valine, N-[(phenylmethoxy)carbonyl]-, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester, GW 327605X, USP Valacyclovir Related Compound E, Ph Eur Valaciclovir Impurity E, Valacyclovir Related Compound E, Valaciclovir Hydrochloride Impurity E, Valaciclovir Impurity E |
| InChI Formula | InChI=1S/C21H26N6O6/c1-13(2)15(24-21(30)33-10-14-6-4-3-5-7-14)19(29)32-9-8-31-12-27-11-23-16-17(27)25-20(22)26-18(16)28/h3-7,11,13,15H,8-10,12H2,1-2H3,(H,24,30)(H3,22,25,26,28)/t15-/m0/s1 |
| SMILES | CC(C)[C@H](NC(=O)OCc1ccccc1)C(=O)OCCOCn2cnc3C(=O)NC(=Nc23)N |
| IUPAC Name | 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?