missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Acetate, Mikromol™

Product Code. 17963136
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963136 100 mg 100mg
17932316 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17963136 Supplier Mikromol Supplier No. MM0061.01
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Acetate
CAS 102728-64-3
Analyte or Component Names 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Acetate
Shipping Condition Room Temperature
Molecular Formula C10 H13 N5 O4
Certifications/Compliance ISO 9001
Synonym Aciclovir Imp. A (EP), Valaciclovir Hydrochloride Imp. I (EP), Valaciclovir Imp. I (EP), Acyclovir USP Related Compound A, Acyclovir USP RC A, 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Acetate, Valaciclovir Hydrochloride Anhydrous Imp. I (EP), 598U75, USP Acyclovir Related Compound A, GW 338914X, Ph Eur Valaciclovir Impurity I, Ph Eur Aciclovir impurity A, Acetylacyclovir, 598U75, Acyclovir Related Compound A, Aciclovir Impurity A, Valaciclovir Hydrochloride Impurity I, Valaciclovir Impurity I
InChI Formula InChI=1S/C10H13N5O4/c1-6(16)19-3-2-18-5-15-4-12-7-8(15)13-10(11)14-9(7)17/h4H,2-3,5H2,1H3,(H3,11,13,14,17)
SMILES CC(=O)OCCOCn1cnc2C(=O)NC(=Nc12)N
IUPAC Name 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl acetate
Molecular Weight (g/mol) 267.24
Quantity 100 mg
Formula Weight 267.0968 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.