missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine, TRC

Product Code. 30004678
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004678 100 mg 100mg
17971885 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004678 Supplier Toronto Research Chemicals Supplier No. TRCA178600100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28910-89-6
Molecular Formula C17 H15 Cl N4 O
Molecular Weight (g/mol) 326.78
Synonym 2-(2-Acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine
IUPAC Name N'-(7-chloro-5-phenyl-3H-1,4-benzodiazepin-2-yl)acetohydrazide
SMILES CC(=O)NNC1=Nc2ccc(Cl)cc2C(=NC1)c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H15ClN4O/c1-11(23)21-22-16-10-19-17(12-5-3-2-4-6-12)14-9-13(18)7-8-15(14)20-16/h2-9H,10H2,1H3,(H,20,22)(H,21,23)
Formula Weight 326.0934
Chemical Name or Material 2-(2-acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.