missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine, Mikromol™

Product Code. 17994346
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994346 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17994346 Supplier Mikromol Supplier No. MM0136.070025
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(2-Acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine
CAS 28910-89-6
Analyte or Component Names 2-(2-acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine
Shipping Condition Room Temperature
Molecular Formula C17 H15 Cl N4 O
Certifications/Compliance ISO 9001
Synonym 2-(2-Acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine, Alprazolam Related Compound A
InChI Formula InChI=1S/C17H15ClN4O/c1-11(23)21-22-16-10-19-17(12-5-3-2-4-6-12)14-9-13(18)7-8-15(14)20-16/h2-9H,10H2,1H3,(H,20,22)(H,21,23)
SMILES CC(=O)NNC1=Nc2ccc(Cl)cc2C(=NC1)c3ccccc3
IUPAC Name N'-(7-chloro-5-phenyl-3H-1,4-benzodiazepin-2-yl)acetohydrazide
Molecular Weight (g/mol) 326.78
Quantity 25 mg
Formula Weight 326.0934 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.