missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[[(2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]-methoxy]ethyl Acetate, Mikromol™
Description
2-[[(2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]-methoxy]ethyl Acetate
Specifications
Specifications
| Chemical Name or Material | 2-[((2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl)-methoxy]ethyl Acetate |
| CAS | 75128-73-3 |
| Analyte or Component Names | 2-[((2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl)-methoxy]ethyl Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12H15N5O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-[[(2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]-methoxy]ethyl Acetate, Aciclovir Imp. G (EP), Aciclovir stage 3, 9-((2-acetoxyethoxy)methyl)-N2-acetylguanine, Acyclovir stage 3, GW 338822X, Ph Eur Aciclovir Impurity G, 808U75 |
| InChI Formula | InChI=1S/C12H15N5O5/c1-7(18)14-12-15-10-9(11(20)16-12)13-5-17(10)6-21-3-4-22-8(2)19/h5H,3-4,6H2,1-2H3,(H2,14,15,16,18,20) |
| SMILES | CC(=O)NC1=Nc2c(ncn2COCCOC(=O)C)C(=O)N1 |
| IUPAC Name | 2-[(2-acetamido-6-oxo-1H-purin-9-yl)methoxy]ethyl acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?