missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethyl-1,3-dioxin-4-one, cont. up to ca 6% acetone

Product Code. 11452387
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452387 500 g 500g
11432387 25 g 25g
11442387 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11452387 Supplier Thermo Scientific Alfa Aesar Supplier No. A14254.36
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 5394-63-8 | C7H10O3 | 142.154 g/mol

2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.

Solubility
Soluble in Chloroform, Methanol. Not miscible in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5394-63-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00040468
InChI Key XFRBXZCBOYNMJP-UHFFFAOYSA-N
Synonym 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb
PubChem CID 79368
IUPAC Name 2,2,6-trimethyl-1,3-dioxin-4-one
SMILES CC1=CC(=O)OC(O1)(C)C

Specifications

Melting Point 7°C
Density 1.094
Boiling Point 65°C to 67°C (2 mmHg)
Flash Point 14°C (57°F)
Refractive Index 1.461
Quantity 500 g
UN Number UN1993
Beilstein 2408
Solubility Information Soluble in Chloroform,Methanol. Not miscible in water.
Formula Weight 142.15
Percent Purity 94%
Assay cont. up to ≈6% acetone
Chemical Name or Material 2,2,6-Trimethyl-1,3-dioxin-4-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.