missing translation for 'onlineSavingsMsg'
Learn More

2-(2,6-Dimethylphenylcarbamoyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 15473938
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473938 1 g 1g
15413436 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473938 Supplier Thermo Scientific Alfa Aesar Supplier No. H57465.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a novel antituberculosis agent.

Solubility
Soluble in (10 mM DMSO).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19368-18-4
Molecular Formula C16H15NO3
Molecular Weight (g/mol) 269.30
MDL Number MFCD00029943
InChI Key LLECMGGNFBKPRH-UHFFFAOYSA-N
Synonym ftaxilide, histanorm, 2',6'-dimethylphthalanilic acid, ftaxilide inn:dcf, ftaxilidum inn-latin, ftaxilida inn-spanish, phthalic 2,6-dimethylanilide, phthalanilic acid, 2',6'-dimethyl, 2-2,6-dimethylphenyl carbamoyl benzoic acid, mp-12-p
PubChem CID 71895
IUPAC Name 2-[(2,6-dimethylphenyl)carbamoyl]benzoic acid
SMILES CC1=CC=CC(C)=C1NC(=O)C1=CC=CC=C1C(O)=O

Specifications

Quantity 1 g
Solubility Information Soluble in (10 mM DMSO).
Formula Weight 269.3
Percent Purity 97%
Chemical Name or Material 2-(2,6-Dimethylphenylcarbamoyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.