missing translation for 'onlineSavingsMsg'
Learn More

2-(2,6-Dichlorophenoxy)-N-phenylacetamide-13C6, TRC

Product Code. 30385750
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385750 10 mg 10mg
30373305 5 mg 5mg
30372805 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385750 Supplier Toronto Research Chemicals Supplier No. TRCD43705410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2,6-Dichlorophenoxy)-N-phenylacetamide-13C6
Molecular Formula 13C6 C8 H11 Cl2 N O2
Quantity 10 mg
InChI Formula InChI=1S/C14H11Cl2NO2/c15-11-7-4-8-12(16)14(11)19-9-13(18)17-10-5-2-1-3-6-10/h1-8H,9H2,(H,17,18)/i1+1,2+1,3+1,5+1,6+1,10+1
SMILES Clc1cccc(Cl)c1OCC(=O)N[13c]2[13cH][13cH][13cH][13cH][13cH]2
IUPAC Name N-((1,2,3,4,5,6-13C6)cyclohexatrienyl)-2-(2,6-dichlorophenoxy)acetamide
Molecular Weight (g/mol) 302.105
Formula Weight 301.037
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.