missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethylpiperidinooxy, 98%

Product Code. 10562221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10562221 25 g Glass bottle 25g
10368240 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10562221 Supplier Thermo Scientific Acros Supplier No. 200380250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2564-83-2 | C9H18NO | 156.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2564-83-2
Molecular Formula C9H18NO
Molecular Weight (g/mol) 156.25
MDL Number MFCD00009599
InChI Key QYTDEUPAUMOIOP-UHFFFAOYSA-N
Synonym TEMPO
IUPAC Name (2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
SMILES CC1(C)CCCC(C)(C)N1[O]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 36°C to 40°C
Color Yellow to Green
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Quantity 25 g
Fieser 16,324; 17,332
Merck Index 15, 9283
Solubility Information Solubility in water: 151mg/l (20°C)
Formula Weight 156.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,2,6,6-Tetramethylpiperidinooxy, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,6,6-Tetramethylpiperidinooxy
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.