missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethylpiperidine, 98%

Product Code. 11926301 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 11926301

Brand: Thermo Scientific Acros 151951000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 768-66-1 | C9H19N | 141.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-66-1
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.26
InChI Key RKMGAJGJIURJSJ-UHFFFAOYSA-N
Synonym norpempidine, piperidine, 2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl piperidine, unii-44n9s1ycfm, 2,2,6,6-tetramethylpeperidine, 44n9s1ycfm, 2,2,6,6 tetramethyl piperidine, 2,2,6,6-tetramethyl-piperidine, pubchem7259, acmc-209p6i
PubChem CID 13035
IUPAC Name 2,2,6,6-tetramethylpiperidine
SMILES CC1(CCCC(N1)(C)C)C

Specifications

Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 152.0°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.4460
Quantity 100 g
Beilstein 20,129
Specific Gravity 0.83
Solubility Information Solubility in water: miscible. Other solubilities: soluble in toluene, dimethylformamide
Formula Weight 141.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,6,6-Tetramethylpiperidine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.