missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2' '-Terpyridine, TRC

Product Code. 30138683
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138683 1 mg 1mg
30136681 2 mg 2mg
30005100 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138683 Supplier Toronto Research Chemicals Supplier No. TRCT1175101MG
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1148-79-4
Molecular Formula C15H11N3
Molecular Weight (g/mol) 233.27
Synonym 2,26,2-Terpyridine Terpy
IUPAC Name 2,6-dipyridin-2-ylpyridine
SMILES c1ccc(nc1)c2cccc(n2)c3ccccn3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C15H11N3/c1-3-10-16-12(6-1)14-8-5-9-15(18-14)13-7-2-4-11-17-13/h1-11H
Formula Weight 233.0953
Chemical Name or Material 2,2':6',2' '-Terpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.