missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine, 97%

Product Code. 11412010
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412010 1 g 1g
11402010 0.25 g 0.25g
11443348 0.25 g 0.25g
15413167 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11412010 Supplier Thermo Scientific Alfa Aesar Supplier No. A17750.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1148-79-4 | C15H11N3 | 233.27 g/mol

2,2';6',2'-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′;6′,2″-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.

Solubility
Soluble in most organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1148-79-4
Molecular Formula C15H11N3
Molecular Weight (g/mol) 233.27
MDL Number MFCD00006213
InChI Key DRGAZIDRYFYHIJ-UHFFFAOYSA-N
Synonym 2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine
PubChem CID 70848
ChEBI CHEBI:245199
IUPAC Name 2,6-dipyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1

Specifications

Melting Point 89°C to 92°C
Assay Percent Range 0.97
Quantity 1 g
UN Number UN2811
Solubility Information Soluble in most organic solvents.
Formula Weight 233.28
Percent Purity 97%
Chemical Name or Material 2, 2':6', 2''-Terpyridine
compliance-icons
Product Identifier
  • 2,2':6',2''-Terpyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,2:6,2-Terpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.