missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine, 96%

Product Code. 15388523 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15388523 250 mg 250mg
15325235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15388523 Supplier Thermo Scientific Acros Supplier No. 215632500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1148-79-4 | C15H11N3 | 233.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1148-79-4
Molecular Formula C15H11N3
Molecular Weight (g/mol) 233.27
MDL Number MFCD00006213
InChI Key DRGAZIDRYFYHIJ-UHFFFAOYSA-N
Synonym 2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine
PubChem CID 70848
ChEBI CHEBI:245199
IUPAC Name 2,6-dipyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1

Specifications

Melting Point 89.0°C to 91.0°C
Boiling Point 370.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Quantity 250 mg
Formula Weight 233.28
Percent Purity 96%
Chemical Name or Material 2,2':6',2''-Terpyridine
compliance-icons
Product Identifier
  • 2,2':6',2''-Terpyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.