missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine-4'-carboxylic acid, 95%

Product Code. 15429105
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429105 250 mg 250mg
15457817 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429105 Supplier Thermo Scientific Alfa Aesar Supplier No. H37760.MD
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 148332-36-9 | C16H11N3O2 | 277.28 g/mol

2,2':6',2″-Terpyridine-4'-carboxylic acid is used as dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′:6′,2′′-Terpyridine-4′-carboxylic acid is used as dyestuff intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148332-36-9
Molecular Formula C16H11N3O2
Molecular Weight (g/mol) 277.28
MDL Number MFCD04114290
InChI Key ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
Synonym 2,2':6',2-terpyridine-4'-carboxylic acid, 2,2':6',2-terpyridine-4'-carboxylicacid, 2,6-bis pyridin-2-yl pyridine-4-carboxylic acid, 6-pyridin-2-yl-2,2'-bipyridine-4-carboxylic acid, acmc-1c3v7, 4'-carboxy-2,2':6',2-terpyridine, 2,2:6,2'-terpyridine-4-carboxylic acid, 1∼2∼,2∼2∼:2∼6∼,3∼2∼-terpyridine-2∼4∼-carboxylic acid, 2,2\\':6\\',2\\'\\'-terpyridine-4\\'-carboxylic acid
PubChem CID 2762749
IUPAC Name 2,6-dipyridin-2-ylpyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 280°C to 282°C
Color White to Cream
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 277.28
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2,2':6',2''-Terpyridine-4'-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.