missing translation for 'onlineSavingsMsg'
Learn More

2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10133964
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10133964 5 g 5g
10780924 250 mg 250mg
10421313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10133964 Supplier Thermo Scientific Maybridge Supplier No. CC39901DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 92028-57-4
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD01125264
InChI Key ZLYUUANOICYAAL-UHFFFAOYSA-N
Synonym 2-2,5-dimethyl-1h-pyrrol-1-yl benzoic acid, 2-2,5-dimethylpyrrol-1-yl benzoic acid, 2-2,5-dimethyl-1-pyrrolyl benzoic acid, 2-2,5-dimethyl-pyrrol-1-yl-benzoic acid, 2-2,5-dimethylpyrrolyl benzoic acid, cbmicro_048042, benzoic acid,2-2,5-dimethyl-1h-pyrrol-1-yl
PubChem CID 292956
IUPAC Name 2-(2,5-dimethylpyrrol-1-yl)benzoic acid
SMILES CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 215.25
Percent Purity 97%
Chemical Name or Material 2-(2, 5-Dimethyl-1H-pyrrol-1-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.