missing translation for 'onlineSavingsMsg'
Learn More

2-(2,5-Dimethoxy-4-methylphenyl)-4,5-dihydro-4,4-dimethyloxazole, TRC

Product Code. 30381997
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381997 50 mg 50mg
30376757 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381997 Supplier Toronto Research Chemicals Supplier No. TRCD49048550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2,5-Dimethoxy-4-methylphenyl)-4,5-dihydro-4,4-dimethyloxazole
CAS 89368-17-2
Molecular Formula C14H19NO3
Quantity 50 mg
Synonym 2-(2,5-Dimethoxy-4-methylphenyl)-4,4-dimethyl-4,5-dihydro-oxazole
InChI Formula InChI=1S/C14H19NO3/c1-9-6-12(17-5)10(7-11(9)16-4)13-15-14(2,3)8-18-13/h6-7H,8H2,1-5H3
SMILES COc1cc(C2=NC(C)(C)CO2)c(OC)cc1C
IUPAC Name 2-(2,5-dimethoxy-4-methylphenyl)-4,4-dimethyl-5H-1,3-oxazole
Molecular Weight (g/mol) 249.31
Formula Weight 249.1365
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.