missing translation for 'onlineSavingsMsg'
Learn More

2,2'-[5-(Bromomethyl)benzene-1,3-diyl]bis(2-methylpropanenitrile), Mikromol™

Product Code. 30051579
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30051579 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 30051579

Brand: Mikromol MM1250.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,2'-[5-(Bromomethyl)benzene-1,3-diyl]bis(2-methylpropanenitrile)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha,alpha,alpha',alpha'-Tetramethyl-5-bromomethyl-1,3-benzenediacetonitrile
CAS 120511-84-4
Analyte or Component Names alpha,alpha,alpha',alpha'-Tetramethyl-5-bromomethyl-1,3-benzenediacetonitrile
Shipping Condition Room Temperature
Molecular Formula C15 H17 Br N2
Certifications/Compliance ISO 9001
Synonym 5-(Bromomethyl)-α1,α1,α3,α3-tetramethyl-1,3-benzenediacetonitrile, 1,3-Benzenediacetonitrile, 5-(bromomethyl)-α,α,α',α'-tetramethyl- (9CI), 5-(Bromomethyl)-α1,α1,α3,α3-tetramethyl-1,3-benzenediacetonitrile, 3,5-Bis(1-cyano-1-methylethyl)benzyl bromide, 3,5-Bis(2-cyanoprop-2-yl)benzyl bromide, 2,2'-[5-(Bromomethyl)benzene-1,3-diyl]bis(2-methylpropanenitrile), Anastrozole Impurity C
InChI Formula InChI=1S/C15H17BrN2/c1-14(2,9-17)12-5-11(8-16)6-13(7-12)15(3,4)10-18/h5-7H,8H2,1-4H3
SMILES CC(C)(C#N)c1cc(CBr)cc(c1)C(C)(C)C#N
IUPAC Name 2-[3-(bromomethyl)-5-(2-cyanopropan-2-yl)phenyl]-2-methylpropanenitrile
Molecular Weight (g/mol) 305.21
Quantity 25 mg
Formula Weight 304.0575 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.