missing translation for 'onlineSavingsMsg'
Learn More

2,2',5,5'-Tetramethylbiphenyl, 98%

Product Code. 11348046
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348046 1 g 1g
11358046 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348046 Supplier Thermo Scientific Alfa Aesar Supplier No. L13330.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3075-84-1 | C16H18 | 210.32 g/mol

2,2',5,5'-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′,5,5′-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3075-84-1
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00151846
InChI Key ZHTROMYSDSTCCE-UHFFFAOYSA-N
PubChem CID 137818
IUPAC Name 2-(2,5-dimethylphenyl)-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(=C1)C1=CC(C)=CC=C1C

Specifications

Melting Point 48°C to 51°C
Quantity 1 g
Beilstein 2046200
Synonym 2,2',5,5'-tetramethylbiphenyl, 2,2',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl, 2,2',5,5'-tetramethyl, biphenyl-, 2,2',5,5'-tetramethyl, 2-2,5-dimethylphenyl-1,4-dimethylbenzene, acmc-20aokf, 2,5,2',5'-tetramethylbiphenyl, 1,1'-biphenyl,2,2',5,5'-tetramethyl, 2,2',5,5'-tetramethyl-1,1'-biphenyl #
Boiling Point 284°C (732 mmHg)
Solubility Information Insoluble in water.
Formula Weight 210.32
Percent Purity 98%
Chemical Name or Material 2,2',5,5'-Tetramethylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.