missing translation for 'onlineSavingsMsg'
Learn More

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide, TRC

Product Code. 30451944
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451944 1 g -
30450306 5 g 5g
30453319 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451944 Supplier Toronto Research Chemicals Supplier No. TRCT3047501G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19805-75-5
Molecular Formula C9 H16 N2 O
Molecular Weight (g/mol) 168.24
Synonym 3-Carbamoyl-2,2,5,5-Tetramethyl-3-Pyrroline
IUPAC Name 2,2,5,5-tetramethyl-1H-pyrrole-3-carboxamide
SMILES CC1(C)NC(C)(C)C(=C1)C(=O)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H16N2O/c1-8(2)5-6(7(10)12)9(3,4)11-8/h5,11H,1-4H3,(H2,10,12)
Formula Weight 168.1263
Chemical Name or Material 2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.