missing translation for 'onlineSavingsMsg'
Learn More

2,2':5',2'-Terthiophene, MedChemExpress

Product Code. 16439176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439176 Supplier Medchem Express Supplier No. HYN2048
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 2,2':5',2'-Terthiophene (α-Terthiophene) is an oligomer of the heterocycle thiophene. 2,2':5',2'-Terthiophene has been employed as building block for the organic semi-conductor polythiophene.

2,2':5',2'-Terthiophene (α-Terthiophene) is an oligomer of the heterocycle thiophene. 2,2':5',2'-Terthiophene has been employed as building block for the organic semi-conductor polythiophene.

Specifications

Chemical Name or Material 2,2':5',2'-Terthiophene
Physical Form Solid
CAS 1081-34-1
Color Green Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H8S3
For Use With (Application) COVID-19-immunoregulation
Synonym α-​Terthiophene α-Terthienyl Trithiophene
Solubility Information DMSO : 100 mg/mL (402.59 mM; Need ultrasonic)
SMILES C1(C2=CC=C(C3=CC=CS3)S2)=CC=CS1
Molecular Weight (g/mol) 248.39
Formula Weight 248.39
Percent Purity 99.55%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.