missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane 95+%, for Residue Analysis, Distol™, Fisher Chemical™
54.50 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Melting Point | -107°C |
Color | Colorless |
Boiling Point | 98°C |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 51mbar at 20°C |
Viscosity | 0.51 mPaS at 22°C |
Formula Weight | 114.23g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'