missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, for HPLC, Fisher Chemical™

Product Code. 10264210 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10264210 5 L Aluminum Can 5 L
10589580 1 L Amber Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10264210 Supplier Fisher Chemical Supplier No. T/3606/21
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

C8H18, CAS Number-540-84-1, 2, 2, 4-trimethylpentane, 2,4,4-trimethylpentane, ch3 2chch2c ch3 3, i-octane, isobutyltrimethylethane, isobutyltrimethylmethane, isooctane, iso-octane, pentane, 2,2,4-trimethyl, unii-qab8f5669o, 2.5L, 98 deg.C, CHEBI:62805, Colorless, 114.23g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number 8943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Color Colorless
Quantity 5 L
Packaging Aluminum Can
Physical Form Liquid
Vapor Pressure 51mbar at 20°C
Viscosity 0.51 mPaS at 22°C
Boiling Point 98°C
Formula Weight 114.23g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.