missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, Environmental Grade, 99.5+%, Thermo Scientific Chemicals

Product Code. 11374877
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for pesticide residue analysis

Suitable for pesticide residue analysis2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Density 0.692 g/mL
Boiling Point 98°C to 99°C
Flash Point -7°C (19°F)
Odor Nearly odorless
Refractive Index 1.391
Quantity 1 L
UN Number UN1262
Beilstein 1696876
Merck Index 14,5193
Solubility Information Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,acetone,heptane,and oils. Immiscible with water.
Formula Weight 114.23
Percent Purity ≥99.5%
Grade Environmental
Physical Form Liquid
Chemical Name or Material 2,2,4-Trimethylpentane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'