missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ACS, 99+%

Product Code. p-4408495
Click to view available options
Quantity:
500 mL
1 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 11400530

Brand: Thermo Scientific Alfa Aesar 031787.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-84-1 | C8H18 | 114.232 g/mol

Solvent, thinner, in determination of fuel octane numbers2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as a thinner. It is involved as a processing aid in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Color Colorless
Density 0.692
Boiling Point 98°C to 99°C
Flash Point -7°C (19°F)
Assay Percent Range >99%
Odor Nearly Odorless
Refractive Index 1.391
Quantity 500 mL
UN Number 1262
Beilstein 1696876
Solubility Information Soluble in benzene,toluene,xylene,chloroform,ether,CS2,CCl4,and DMF. Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,acetone,heptane and oils. Immiscible with water.
Formula Weight 114.23
Vapor Pressure 15 hPa (11mm Hg) at 20°C
Grade ACS
Physical Form Liquid
Chemical Name or Material 2, 2, 4-Trimethylpentane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.