missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ACS, 99+%

Codice prodotto. 11348287
Cambia vista
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Dimensione della confezione:
1 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
11348287 4 L 4 L
11400530 500 mL 500mL
11328287 1 L 1 L
11338287 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11348287 Fornitore Thermo Scientific Alfa Aesar N. del fornitore 031787.K7
 Restricted Item
Estimated Shipment: 21-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 540-84-1 | C8H18 | 114.232 g/mol

Solvent, thinner, in determination of fuel octane numbers2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as a thinner. It is involved as a processing aid in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifica

Melting Point -107C
Color Colorless
Density 0.692 g/cm3 at 20°C
Boiling Point 98°C to 99°C
Flash Point -7°C (19°F)
Assay Percent Range >99%
Odor Nearly Odorless
Refractive Index 1.391
Quantity 4 L
UN Number 1262
Beilstein 1696876
Solubility Information Soluble in benzene,toluene,xylene,chloroform,ether,CS2,CCl4,and DMF. Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,acetone,heptane and oils. Immiscible with water.
Formula Weight 114.23
Vapor Pressure 15 hPa (11mm Hg) at 20°C
Grade ACS
Physical Form Liquid
Chemical Name or Material 2, 2, 4-Trimethylpentane
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.