Product Code. 15905620
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
15905620
7 L
7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
15905620
Supplier
Solstice
Supplier No.
322917L
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
2,2,4-Trimethylpentane, Puriss. p.a., ≥99.5% (GC).
Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals. It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
pH
No data available
Melting Point
-107°C
CAS
540-84-1
Color
Colorless
Density
0.69 g/cm3 (20°C)
Quantity
7 L
Flash Point
-12°C
Packaging
Returnable Stainless Steel Can
Physical Form
Liquid
Product Line
Puriss. p.a.
Molecular Formula
C8H18
Linear Formula
(CH3 )2 CHCH2 C(CH3 )3
Vapor Pressure
55 hPa (21°C)
MDL Number
MFCD00008943
Viscosity
No data available
UN Number
UN1262
Beilstein
1696876
Synonym
Isooctane
Boiling Point
99°C (1.013 hPa)
InChI Key
NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES
CC(C)CC(C)(C)C
IUPAC Name
2,2,4-trimethylpentane
Molecular Weight (g/mol)
114.23
Percent Purity
≥99.50%
Grade
ACS
Chemical Name or Material
2,2,4-Trimethylpentane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?